Methyl-4-benzyloxybenzoate
Dodavatel: Apollo Scientific
APOSOR111495-1GEA
1580
CZK
APOSOR111495-1G
Methyl-4-benzyloxybenzoate
Methyl-4-benzyloxybenzoate
Vzorec:
C₁₅H₁₄O₃ MW: 242,27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00017613 CAS číslo: 32122-11-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...