Tisk…

5-Carboxyphthalide 97%

Dodavatel: Apollo Scientific

Varování

APOSOR11405-1GEA 5160 CZK
APOSOR11405-1G
5-Carboxyphthalide 97%
5-Carboxyphthalide
Vzorec: C₉H₆O₄
MW: 178,1400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04117971
CAS číslo: 4792-29-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR