3-Chloro-6-(pyrrolidin-1-yl)pyridazine 97%
APOSOR11425-5GEA
0
CZK
APOSOR11425-5G
APOSOR11425-1G
3-Chloro-6-(pyrrolidin-1-yl)pyridazine 97%
3-Chloro-6-(pyrrolidin-1-yl)pyridazine
Vzorec:
C₈H₁₀ClN₃ Storage Temperature: Ambient |
MDL Number:
MFCD05864735 CAS číslo: 66346-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...