Tisk…

3-Chloro-4-(cyclohexylcarbamoyl)phenylboronic acid 96%

Dodavatel: Apollo Scientific

APOSOR11445-1GEA 5200 CZK
APOSOR11445-1G
3-Chloro-4-(cyclohexylcarbamoyl)phenylboronic acid 96%
3-Chloro-4-(cyclohexylcarbamoyl)phenylboronic acid
Vzorec: C₁₃H₁₇BClNO₃
MW: 281,5400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09475818
CAS číslo: 957034-65-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR