Tisk…

6-Bromo-3-(methylthio)[1,2,4]triazolo[4,3-a]pyridine 98%

Dodavatel: Apollo Scientific

APOSOR11485-250MGEA 8100 CZK
APOSOR11485-250MG APOSOR11485-1G
6-Bromo-3-(methylthio)[1,2,4]triazolo[4,3-a]pyridine 98%
6-Bromo-3-(methylthio)[1,2,4]triazolo[4,3-a]pyridine
Vzorec: C₇H₆BrN₃S
Storage Temperature: Refrigerator
MDL Number: MFCD09475863
CAS číslo: 941294-56-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR