Tisk…

3-Bromo-N-4-dimethylbenzenesulphonamide 98%

APOSOR11526-1GEA 0 CZK
APOSOR11526-1G APOSOR11526-5G
3-Bromo-N-4-dimethylbenzenesulphonamide 98%
3-Bromo-N-4-dimethylbenzenesulphonamide
Vzorec: C₈H₁₀BrNO₂S
Storage Temperature: Ambient
MDL Number: MFCD09475907
CAS číslo: 941294-51-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR