Tisk…

5-Methylimidazo[1,2-a]pyridine hydrochloride 98%

Varování

APOSOR11553-1GEA 0 CZK
APOSOR11553-1G APOSOR11553-5G
5-Methylimidazo[1,2-a]pyridine hydrochloride 98%
5-Methylimidazo[1,2-a]pyridine hydrochloride
Vzorec: C₈H₉ClN₂
Storage Temperature: Refrigerator
MDL Number: MFCD09475894
CAS číslo: 5857-49-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR