Tisk…

2-Bromo-N-methylaniline hydrochloride

APOSOR11598-5GEA 0 CZK
APOSOR11598-5G APOSOR11598-1G
2-Bromo-N-methylaniline hydrochloride
2-Bromo-N-methylaniline hydrochloride
Vzorec: C₇H₉BrClN
Storage Temperature: Ambient
MDL Number: MFCD09800853
CAS číslo: 957120-69-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR