Tisk…

5-Bromo-4-p-tolylpyrimidine

Dodavatel: Apollo Scientific

APOSOR11614-5GEA 11610 CZK
APOSOR11614-5G APOSOR11614-1G
5-Bromo-4-p-tolylpyrimidine
5-Bromo-4-p-tolylpyrimidine
Vzorec: C₁₂H₁₁BrN₂
Storage Temperature: Ambient
MDL Number: MFCD09800948
CAS číslo: 149323-50-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR