Tisk…

4-Bromo-N-cyclohexylbenzamide

APOSOR11621-1GEA 0 CZK
APOSOR11621-1G APOSOR11621-5G
4-Bromo-N-cyclohexylbenzamide
4-Bromo-N-cyclohexylbenzamide
Vzorec: C₁₃H₁₆BrNO
Storage Temperature: Ambient
MDL Number: MFCD00451750
CAS číslo: 223553-87-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR