Tisk…

2-Bromo-N-tert-butylbenzenesulphonamide

Dodavatel: Apollo Scientific

APOSOR11648-1GEA 3740 CZK
APOSOR11648-1G APOSOR11648-5G
2-Bromo-N-tert-butylbenzenesulphonamide
2-Bromo-N-tert-butylbenzenesulphonamide
Vzorec: C₁₀H₁₄BrNO₂S
Storage Temperature: Ambient
MDL Number: MFCD07710752
CAS číslo: 138733-50-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR