Tisk…

3-Bromo-N-(2-hydroxyethyl)benzamide

Dodavatel: Apollo Scientific

Varování

APOSOR11911-5GEA 14520 CZK
APOSOR11911-5G APOSOR11911-1G
3-Bromo-N-(2-hydroxyethyl)benzamide
3-Bromo-N-(2-hydroxyethyl)benzamide
Vzorec: C₉H₁₀BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD01249209
CAS číslo: 57728-66-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR