2-Methoxy-4-methylaniline hydrochloride
|
Varování
|
APOSOR11969-1GEA
0
CZK
APOSOR11969-1G
2-Methoxy-4-methylaniline hydrochloride
2-Methoxy-4-methylaniline hydrochloride
Vzorec:
C₈H₁₂ClNO MW: 173,6400 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09878311 CAS číslo: 71288-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...