Tisk…

Methyl-4-chloropicolinate hydrochloride

Varování

APOSOR1204-25GEA 0 CZK
APOSOR1204-25G APOSOR1204-5G
Methyl-4-chloropicolinate hydrochloride
Methyl-4-chloropicolinate hydrochloride
Vzorec: C₇H₇Cl₂NO₂
Bod tání: 136…138 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05864729
CAS číslo: 176977-85-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR