2-((4-Bromophenyl)amino)acetophenone
Dodavatel: Apollo Scientific
APOSOR12171-10GEA
7210
CZK
APOSOR12171-10G
APOSOR12171-1G
APOSOR12171-5G
2-((4-Bromophenyl)amino)acetophenone
2-((4-Bromophenyl)amino)acetophenone
Vzorec:
C₁₄H₁₂BrNO Bod tání: 158…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00598456 CAS číslo: 4831-21-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...