(2,4-Dichlorophenoxy)thioacetamide
Dodavatel: Apollo Scientific
APOSOR12187-10GEA
10950
CZK
APOSOR12187-10G
APOSOR12187-1G
APOSOR12187-5G
(2,4-Dichlorophenoxy)thioacetamide
(2,4-Dichlorophenoxy)thioacetamide
Vzorec:
C₈H₇Cl₂NOS MW: 236,11 g/mol Bod tání: 139…141 °C Storage Temperature: Ambient |
MDL Number:
MFCD00172128 CAS číslo: 2302-32-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...