3-Chloro-4-ethoxyphenylboronic acid 98%
|
Varování
|
APOSOR13156-25GEA
0
CZK
APOSOR13156-25G
APOSOR13156-5G
APOSOR13156-10G
3-Chloro-4-ethoxyphenylboronic acid 98%
3-Chloro-4-ethoxyphenylboronic acid
Vzorec:
C₈H₁₀BClO₃ MW: 200,4300 g/mol Bod tání: 230…236 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03701556 CAS číslo: 279261-81-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...