Tisk…

2-Amino-6-chloro-4-nitrophenol 97%

Dodavatel: Apollo Scientific

APOSOR13236-25GEA 3940 CZK
APOSOR13236-25G
2-Amino-6-chloro-4-nitrophenol 97%
2-Amino-6-chloro-4-nitrophenol
Vzorec: C₆H₅ClN₂O₃
MW: 188,57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00035767
CAS číslo: 6358-09-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR