Ethyl-2-amino-4-oxazolecarboxylate
Dodavatel: Apollo Scientific
|
Varování
|
APOSOR1342-1GEA
457
CZK
APOSOR1342-1G
Ethyl-2-amino-4-oxazolecarboxylate
Ethyl-2-amino-4-oxazolecarboxylate
Vzorec:
C₆H₈N₂O₃ MW: 156,14 g/mol Bod tání: 124…128 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06659907 CAS číslo: 177760-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...