3-Pyridinesulphonic acid 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
APOSOR13637-100GEA
2990
CZK
APOSOR13637-100G
3-Pyridinesulphonic acid 97%
3-Pyridinesulphonic acid
Vzorec:
C₅H₅NO₃S MW: 159,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00006381 CAS číslo: 636-73-7 UN: 2585 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...