Order Entry
3-(1H-Pyrazol-3-yl)benzoic acid 97%
Irritant
3-(1H-Pyrazol-3-yl)benzoic acid 97%
Supplier:  Apollo Scientific
Certificates
Irritant
3-(1H-Pyrazol-3-yl)benzoic acid 97%
Supplier:  Apollo Scientific
Product Information
  • Catalog #
    Supplier #
    Pack type
    Pk
    Availability
    Price
  • APOSOR1400-250MG
    OR1400-250MG
    Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 1 240,00 Kč
    1 240,00 Kč
    1 * 250 mg
       1 240,00 Kč
    Supplier Article Number: OR1400-250MG
    Local Article Number: APOSOR1400-250MG
  • APOSOR1400-1G
    OR1400-1G
    Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 4 370,00 Kč
    4 370,00 Kč
    1 * 1 g
       4 370,00 Kč
    Supplier Article Number: OR1400-1G
    Local Article Number: APOSOR1400-1G