Tisk…

2-Hydrazino-6-methylbenzothiazole

APOSOR14131-5GEA 0 CZK
APOSOR14131-5G APOSOR14131-1G APOSOR14131-10G
2-Hydrazino-6-methylbenzothiazole
2-Hydrazino-6-methylbenzothiazole
Vzorec: C₈H₉N₃S
Bod tání: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD04448800
CAS číslo: 20174-69-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR