(S)-2-Amino-4-phenylbutanoic acid 98%
Dodavatel: Apollo Scientific
APOSOR14697-100GEA
2610
CZK
APOSOR14697-100G
(S)-2-Amino-4-phenylbutanoic acid 98%
(S)-2-Amino-4-phenylbutanoic acid
Vzorec:
C₁₀H₁₃NO₂ MW: 179,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002619 CAS číslo: 943-73-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...