Tisk…

6-Amino-2-(4-bromophenyl)imidazo[1,2-a]pyridine

Dodavatel: Apollo Scientific

APOSOR15317-500MGEA 11380 CZK
APOSOR15317-500MG
6-Amino-2-(4-bromophenyl)imidazo[1,2-a]pyridine
6-Amino-2-(4-bromophenyl)imidazo[1,2-a]pyridine
Vzorec: C₁₃H₁₀BrN₃
Bod tání: 191…195 °C
Storage Temperature: Ambient
MDL Number: MFCD08276939
CAS číslo: 885950-52-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR