3-Phenyl-5-isoxazolecarboxylic acid 97%
|
Varování
|
APOSOR15371-5GEA
0
CZK
APOSOR15371-5G
APOSOR15371-1G
3-Phenyl-5-isoxazolecarboxylic acid 97%
3-Phenyl-5-isoxazolecarboxylic acid
Vzorec:
C₁₀H₇NO₃ MW: 189,17 g/mol Bod tání: 178…179,5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01936010 CAS číslo: 14442-12-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...