Tisk…

2-(Methylthio)-4-(tributylstannyl)pyrimidine

Nebezpečí

APOSOR15529-5GEA 22580 CZK
APOSOR15529-5G APOSOR15529-1G
2-(Methylthio)-4-(tributylstannyl)pyrimidine
2-(Methylthio)-4-(tributylstannyl)pyrimidine
Vzorec: C₁₇H₃₂N₂SSn
Storage Temperature: Ambient
MDL Number: MFCD01319026
CAS číslo: 123061-49-4
UN: 2788
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR