5-Methyl-2-(tributylstannyl)thiazole 96%
|
Nebezpečí
|
APOSOR15545-250MGEA
4570
CZK
APOSOR15545-250MG
APOSOR15545-1G
5-Methyl-2-(tributylstannyl)thiazole 96%
5-Methyl-2-(tributylstannyl)thiazole
Vzorec:
C₁₆H₃₁NSSn MW: 388,19 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09025809 CAS číslo: 848613-91-2 UN: 2788 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...