Tisk…

5-Amino-8-chloroquinoline

Dodavatel: Apollo Scientific

APOSOR16165-1GEA 14920 CZK
APOSOR16165-1G APOSOR16165-250MG
5-Amino-8-chloroquinoline
5-Amino-8-chloroquinoline
Vzorec: C₉H₇ClN₂
Bod tání: 147…149 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09802113
CAS číslo: 75793-58-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR