(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid 95+%
Dodavatel: Apollo Scientific
|
Varování
|
APOSOR17020-100GEA
1990
CZK
APOSOR17020-100G
(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid 95+%
(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid
Vzorec:
C₁₇H₁₇NO₄ MW: 299,3 g/mol Bod tání: 85…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020418 CAS číslo: 1161-13-3 EINECS: 214-599-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...