3-(Cyanomethoxy)phenylboronic acid 95%
APOSOR17814-250MGEA
0
CZK
APOSOR17814-250MG
APOSOR17814-1G
3-(Cyanomethoxy)phenylboronic acid 95%
3-(Cyanomethoxy)phenylboronic acid
Vzorec:
C₈H₈BNO₃ MW: 176,9700 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09908169 CAS číslo: 947533-25-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...