Tisk…

3-(2,5-Dimethylphenylcarbamoyl)phenylboronic acid 98%

Varování

APOSOR2098-1GEA 0 CZK
APOSOR2098-250MG APOSOR2098-1G APOSOR2098-5G APOSOR2098-25G
3-(2,5-Dimethylphenylcarbamoyl)phenylboronic acid 98%
3-(2,5-Dimethylphenylcarbamoyl)phenylboronic acid
Vzorec: C₁₅H₁₆BNO₃
Storage Temperature: Ambient
MDL Number: MFCD09027263
CAS číslo: 957061-00-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR