Tisk…

1-(2-Cyanophenyl)piperidine

Dodavatel: Apollo Scientific

APOSOR21258-25GEA 7010 CZK
APOSOR21258-25G
1-(2-Cyanophenyl)piperidine
1-(2-Cyanophenyl)piperidine
Vzorec: C₁₂H₁₄N₂
MW: 186,26 g/mol
Bod varu: 122…125 °C/1mmHg
Bod tání: 41…45 °C
Storage Temperature: Ambient
MDL Number: MFCD00049221
CAS číslo: 72752-52-4
EINECS: 427-330-1
UN: 3077
ADR: 9+EHS,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR