2-Methyl-4-thiazolecarboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
APOSOR23080-5GEA
0
CZK
APOSOR23080-5G
APOSOR23080-250MG
APOSOR23080-1G
2-Methyl-4-thiazolecarboxylic acid
2-Methyl-4-thiazolecarboxylic acid
Vzorec:
C₅H₅NO₂S MW: 143,1637 g/mol Bod tání: 145…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD03407332 CAS číslo: 35272-15-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...