4-(Thiophen-2-yl)benzoic acid
Dodavatel: Apollo Scientific
APOSOR23163-5GEA
4170
CZK
APOSOR23163-5G
4-(Thiophen-2-yl)benzoic acid
4-(Thiophen-2-yl)benzoic acid
Vzorec:
C₁₁H₈O₂S MW: 204,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00690742 CAS číslo: 29886-62-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...