(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide 97%
APOSOR24913-100GEA
0
CZK
APOSOR24913-100G
(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide 97%
(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide
Vzorec:
C₂₂H₂₂BrO₂P MW: 429,30 g/mol Bod tání: 193…195 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011966 CAS číslo: 52509-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...