(4-Methylphenylthio)acetone
Dodavatel: Apollo Scientific
|
Varování
|
APOSOR26023-1GEA
500
CZK
APOSOR26023-1G
(4-Methylphenylthio)acetone
(4-Methylphenylthio)acetone
Vzorec:
C₁₀H₁₂OS MW: 180,27 g/mol Bod varu: 96…98 °C/0,1mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00052713 CAS číslo: 1200-13-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...