Tisk…

4-Amino-6-hydroxy-2-mercaptopyrimidine hydrate

Dodavatel: Apollo Scientific
Varování

APOSOR2651-5GEA 417 CZK
APOSOR2651-5G
4-Amino-6-hydroxy-2-mercaptopyrimidine hydrate
4-Amino-6-hydroxy-2-mercaptopyrimidine hydrate
Vzorec: C₄H₅N₃OS·xH₂O
MW: 143,17 g/mol (anhy)
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00052384
CAS číslo: 65802-56-4
EINECS: 213-722-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR