3-Pyridyloxyacetic acid 98+%
Dodavatel: Apollo Scientific
|
Varování
|
APOSOR300774-1GEA
7010
CZK
APOSOR300774-1G
3-Pyridyloxyacetic acid 98+%
3-Pyridyloxyacetic acid
Vzorec:
C₇H₇NO₃ MW: 153,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04039772 CAS číslo: 86649-57-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...