Tisk…

3-Amino-5-methoxypyridine 97%

Dodavatel: Apollo Scientific

Varování

APOSOR301042-25GEA 15840 CZK
APOSOR301042-25G APOSOR301042-5G APOSOR301042-1G
3-Amino-5-methoxypyridine 97%
3-Amino-5-methoxypyridine
Vzorec: C₆H₈N₂O
MW: 124,1400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09701369
CAS číslo: 64436-92-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR