Tisk…

Pyrrolidine-3-carboxylic acid

Dodavatel: Apollo Scientific

APOSOR301045-10GEA 13090 CZK
APOSOR301045-10G
Pyrrolidine-3-carboxylic acid
Pyrrolidine-3-carboxylic acid
Vzorec: C₅H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD01318699
CAS číslo: 59378-87-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR