N1-Benzylpropane-1,3-diamine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
APOSOR301059-1GEA
1660
CZK
APOSOR301059-1G
N1-Benzylpropane-1,3-diamine
N1-Benzylpropane-1,3-diamine
Vzorec:
C₁₀H₁₆N₂ MW: 164,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00274361 CAS číslo: 13910-48-0 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...