(1H-Indol-2-yl)methanamine
Dodavatel: Apollo Scientific
APOSOR301148-1GEA
3340
CZK
APOSOR301148-1G
(1H-Indol-2-yl)methanamine
(1H-Indol-2-yl)methanamine
Vzorec:
C₉H₁₀N₂ MW: 146,19 g/mol Storage Temperature: Ambient |
CAS číslo:
21109-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...