Methyl-3-chloro-4-methylbenzoate
|
Varování
|
APOSOR30242-1GEA
0
CZK
APOSOR30242-1G
APOSOR30242-25G
APOSOR30242-5G
Methyl-3-chloro-4-methylbenzoate
Methyl-3-chloro-4-methylbenzoate
Vzorec:
C₉H₉ClO₂ MW: 184,62 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00205141 CAS číslo: 56525-63-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...