Tisk…

4-Bromopyrimidine-2-carbonitrile

Dodavatel: Apollo Scientific

Varování

APOSOR302429-1GEA 26520 CZK
APOSOR302429-1G
4-Bromopyrimidine-2-carbonitrile
4-Bromopyrimidine-2-carbonitrile
Vzorec: C₅H₂BrN₃
Storage Temperature: Ambient
MDL Number: MFCD12400798
CAS číslo: 1160995-48-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR