Tisk…

Methyl-2-amino-3-chlorobenzoate

Dodavatel: Apollo Scientific

APOSOR302558-1GEA 15840 CZK
APOSOR302558-1G
Methyl-2-amino-3-chlorobenzoate
Methyl-2-amino-3-chlorobenzoate
Vzorec: C₈H₈ClNO₂
MW: 185,6100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06797833
CAS číslo: 77820-58-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR