4-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester
APOSOR303401-500MGEA
0
CZK
APOSOR303401-500MG
APOSOR303401-1G
APOSOR303401-5G
4-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester
4-(4-Morpholinylcarbonyl)phenylboronic acid pinacol ester
Vzorec:
C₁₇H₂₄BNO₄ MW: 317,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05863913 CAS číslo: 656239-38-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...