4-(3-(Ethoxycarbonyl)piperidine-1-carbonyl)phenylboronic acid pinacol ester
Dodavatel: Apollo Scientific
APOSOR303404-5GEA
7480
CZK
APOSOR303404-5G
APOSOR303404-1G
APOSOR303404-500MG
4-(3-(Ethoxycarbonyl)piperidine-1-carbonyl)phenylboronic acid pinacol ester
4-(3-(Ethoxycarbonyl)piperidine-1-carbonyl)phenylboronic acid pinacol ester
Vzorec:
C₂₁H₃₀BNO₅ MW: 387,28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06657751 CAS číslo: 850411-14-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...