Tisk…

4-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester

APOSOR303407-1GEA 0 CZK
APOSOR303407-1G APOSOR303407-500MG APOSOR303407-5G
4-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester
4-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester
Vzorec: C₁₈H₂₇BN₂O₃
MW: 330,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05863914
CAS číslo: 832114-06-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR