Tisk…

5-(Methoxycarbonyl)-2-methylphenylboronic acid pinacol ester

Dodavatel: Apollo Scientific

APOSOR303533-500MGEA 5890 CZK
APOSOR303533-500MG APOSOR303533-1G
5-(Methoxycarbonyl)-2-methylphenylboronic acid pinacol ester
5-(Methoxycarbonyl)-2-methylphenylboronic acid pinacol ester
Vzorec: C₁₅H₂₁BO₄
MW: 276,1358 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520530
CAS číslo: 882679-40-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR