5-Bromo-2-chloronicotinamide
APOSOR303623-1GEA
0
CZK
APOSOR303623-1G
APOSOR303623-500MG
5-Bromo-2-chloronicotinamide
5-Bromo-2-chloronicotinamide
Vzorec:
C₆H₄BrClN₂O MW: 235,4658 g/mol Bod tání: 184…186 °C Storage Temperature: Ambient |
MDL Number:
MFCD13176452 CAS číslo: 75291-85-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...